Thermo Scientific Chemicals

Sodium perborate monohydrate, 95%, Thermo Scientific Chemicals

Catalog number: 386410250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium perborate monohydrate, 95%, Thermo Scientific Chemicals

Catalog number: 386410250
25 g, Plastic bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 386410250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10332-33-9
IUPAC Name
sodium hydrate [(oxoboranyl)oxy]olate
Molecular Formula
BH2NaO4
InChI Key
MDGXUEVTGARGDK-UHFFFAOYSA-M
SMILES
O.[Na+].[O-]OB=O
Appearance (Color)
White
Appearance (Form)
Crystalline or granular powder
Titration with KMnO4
15 to 16 % (active oxygen)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text