trans-beta-Methyl-beta-nitrostyrene, 99%, Thermo Scientific Chemicals
trans-beta-Methyl-beta-nitrostyrene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-beta-Methyl-beta-nitrostyrene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 386510050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS705-60-2
IUPAC Name[(1E)-2-nitroprop-1-en-1-yl]benzene
Molecular FormulaC9H9NO2
InChI KeyWGSVFWFSJDAYBM-BQYQJAHWSA-N
SMILESCC(=C/C1=CC=CC=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point64°C to 67°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only