Thermo Scientific Chemicals

tert-Butyl (S)-(-)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%, Thermo Scientific Chemicals

Catalog number: 386570010
1 g, Each
Thermo Scientific Chemicals

tert-Butyl (S)-(-)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%, Thermo Scientific Chemicals

Catalog number: 386570010
1 g, Each
Quantity
Catalog number: 386570010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
102308-32-7
IUPAC Name
tert-butyl (4S)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
Molecular Formula
C11H19NO4
InChI Key
PNJXYVJNOCLJLJ-MRVPVSSYSA-N
SMILES
CC(C)(C)OC(=O)N1[C@@H](COC1(C)C)C=O
Appearance (Color)
Clear colorless to yellow
GC
>=94 %
Infrared spectrum
Conforms
Specific optical rotation
-85° to -95° (20°C, 589 nm) (c=1, CHCl3)
Appearance (Form)
Viscous liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text