Thermo Scientific Chemicals

Choline hydroxide, 45 wt.% aqueous solution, pure, stabilized

Catalog number: 386721000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Choline hydroxide, 45 wt.% aqueous solution, pure, stabilized

Catalog number: 386721000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 386721000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-41-1, 7732-18-5
Appearance (Color)
Colorless to tan
Stabilizer
about 0.2 % ethylenediamine
Appearance (Form)
Viscous solution
Titration with HClO4
44.0 to 48.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text