Perfluorooctyl bromide, 99%, Thermo Scientific Chemicals
Perfluorooctyl bromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Perfluorooctyl bromide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 386790500
Price (USD)/ Each
458.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
539.00 
Save 80.35 (15%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
458.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
539.00 
Save 80.35 (15%)
Add to cart
Perfluorooctyl bromide, 99%, Thermo Scientific Chemicals
Catalog number386790500
Price (USD)/ Each
458.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
539.00 
Save 80.35 (15%)
-
Add to cart
Chemical Identifiers
CAS423-55-2
IUPAC Name1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane
Molecular FormulaC8BrF17
InChI KeyWTWWXOGTJWMJHI-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only