Thermo Scientific Chemicals

Tricyclohexylphosphonium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: 386830010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Tricyclohexylphosphonium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: 386830010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 386830010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58656-04-5
IUPAC Name
tetrafluoroboranuide; tricyclohexylphosphanium
Molecular Formula
C18H34BF4P
InChI Key
MYSMMEUXKHJYKH-UHFFFAOYSA-O
SMILES
F[B-](F)(F)F.C1CCC(CC1)[PH+](C1CCCCC1)C1CCCCC1
Appearance (Form)
Powder
Infrared spectrum
Conforms
Elemental Analysis
>=98.5 %
Appearance (Color)
White
Melting point
165°C to 169°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text