Thermo Scientific Chemicals

Di-tert-butylmethylphosphonium tetrafluoroborate 99%, Thermo Scientific Chemicals

Catalog number: 386960050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Di-tert-butylmethylphosphonium tetrafluoroborate 99%, Thermo Scientific Chemicals

Catalog number: 386960050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 386960050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
479094-62-7
IUPAC Name
di-tert-butyl(methyl)phosphanium; tetrafluoroboranuide
Molecular Formula
C9H22BF4P
InChI Key
BRDLRXCAHKUWJS-UHFFFAOYSA-O
SMILES
F[B-](F)(F)F.C[PH+](C(C)(C)C)C(C)(C)C
Infrared spectrum
Conforms
NMR (31P-NMR)
>=99
NMR (1H-NMR)
Conforms
Appearance (Form)
Crystalline powder
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text