2-Octyn-1-ol, 98%, Thermo Scientific Chemicals
2-Octyn-1-ol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Octyn-1-ol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number 387030250
Price (USD)/ Each
135.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
159.00 
Save 23.35 (15%)
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
135.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
159.00 
Save 23.35 (15%)
Add to cart
2-Octyn-1-ol, 98%, Thermo Scientific Chemicals
Catalog number387030250
Price (USD)/ Each
135.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
159.00 
Save 23.35 (15%)
-
Add to cart
Chemical Identifiers
CAS20739-58-6
IUPAC Nameoct-2-yn-1-ol
Molecular FormulaC8H14O
InChI KeyTTWYFVOMGMBZCF-UHFFFAOYSA-N
SMILESCCCCCC#CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4550 to 1.4570 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only