Sodium sulfide nonahydrate, 98+%, extra pure, Thermo Scientific Chemicals
Sodium sulfide nonahydrate, 98+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium sulfide nonahydrate, 98+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 387060025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namedisodium nonahydrate sulfanediide
Molecular FormulaH18Na2O9S
InChI KeyZGHLCBJZQLNUAZ-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Molecular Weight (g/mol)240.17
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to light yellow
Appearance (Form)Powder or crystals and/or chunks
Titration Iodimetric32.0 to 38.0 % (Na2S)
Ammonium (NH4)=<0.005 %
Sulfite (as SO2)=<2000 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only