Thermo Scientific Chemicals

N-Vinylcarbazole, 97%, Thermo Scientific Chemicals

Catalog number: 387221000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Vinylcarbazole, 97%, Thermo Scientific Chemicals

Catalog number: 387221000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 387221000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1484-13-5
IUPAC Name
9-ethenyl-9H-carbazole
Molecular Formula
C14H11N
InChI Key
KKFHAJHLJHVUDM-UHFFFAOYSA-N
SMILES
C=CN1C2=C(C=CC=C2)C2=C1C=CC=C2
Appearance (Color)
Off-white to yellow
Appearance (Form)
Crystalline powder
Melting point
62°C to 66°C
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text