Thermo Scientific Chemicals

Cellulose microcrystalline, average particle size 50μm, Thermo Scientific Chemicals

Catalog number: 387240010
1 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Cellulose microcrystalline, average particle size 50μm, Thermo Scientific Chemicals

Catalog number: 387240010
1 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 387240010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-34-6
IUPAC Name
2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol
Molecular Formula
C14H26O11
InChI Key
PTHCMJGKKRQCBF-UHFFFAOYNA-N
SMILES
COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O
Particle size
>=50 % (> 32 µm)
Appearance (Color)
White
Infrared spectrum
Conforms
Heavy metals (as Pb)
=<10 ppm
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text