Chemical Identifiers
CAS9004-34-6
IUPAC Name2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol
Molecular FormulaC14H26O11
InChI KeyPTHCMJGKKRQCBF-UHFFFAOYNA-N
SMILESCOC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O
View more