Thermo Scientific Chemicals

Lead(II) sulfate, 99%, Thermo Scientific Chemicals

Catalog number: 387275000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Lead(II) sulfate, 99%, Thermo Scientific Chemicals

Catalog number: 387275000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 387275000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7446-14-2
IUPAC Name
λ²-lead(2+) sulfate
Molecular Formula
O4PbS
InChI Key
KEQXNNJHMWSZHK-UHFFFAOYSA-L
SMILES
[Pb++].[O-]S([O-])(=O)=O
Appearance (Form)
Crystalline powder
Identification
Confirmed
Appearance (Color)
White
Elemental Analysis
>=98.5 % (AES)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text