4,4'-Biphenyldicarboxylic acid, 98%, Thermo Scientific Chemicals
4,4'-Biphenyldicarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,4'-Biphenyldicarboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 387300250
Price (USD)/ Each
243.65
Online exclusive
271.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
243.65
Online exclusive
271.00 
Save 27.35 (10%)
Add to cart
4,4'-Biphenyldicarboxylic acid, 98%, Thermo Scientific Chemicals
Catalog number387300250
Price (USD)/ Each
243.65
Online exclusive
271.00 
Save 27.35 (10%)
-
Add to cart
Chemical Identifiers
CAS787-70-2
IUPAC Name[1,1'-biphenyl]-4,4'-dicarboxylic acid
Molecular FormulaC14H10O4
InChI KeyNEQFBGHQPUXOFH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
HPLC>=97.5 %
Appearance (Color)White to light beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only