Thermo Scientific Chemicals

Nickel(II) chloride, 98%, Thermo Scientific Chemicals

Catalog number: 387320500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Nickel(II) chloride, 98%, Thermo Scientific Chemicals

Catalog number: 387320500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 387320500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7718-54-9
IUPAC Name
nickel(2+) dichloride
Molecular Formula
Cl2Ni
InChI Key
QMMRZOWCJAIUJA-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Ni++]
Appearance (Color)
Light orange to brown
Titration Argentometric
>=97.5 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text