Benzyl dimethyl phosphonoacetate, 98%, Thermo Scientific Chemicals
Benzyl dimethyl phosphonoacetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyl dimethyl phosphonoacetate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 387370050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS57443-18-2
IUPAC Namebenzyl 2-(dimethoxyphosphoryl)acetate
Molecular FormulaC11H15O5P
InChI KeyQYLGNJMIOVHLQQ-UHFFFAOYSA-N
SMILESCOP(=O)(CC(=O)OCC1=CC=CC=C1)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light brown
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5050 to 1.5070 (20°C, 589 nm)
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only