Thermo Scientific Chemicals

Benzyl dimethyl phosphonoacetate, 98%, Thermo Scientific Chemicals

Catalog number: 387370250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Benzyl dimethyl phosphonoacetate, 98%, Thermo Scientific Chemicals

Catalog number: 387370250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 387370250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57443-18-2
IUPAC Name
benzyl 2-(dimethoxyphosphoryl)acetate
Molecular Formula
C11H15O5P
InChI Key
QYLGNJMIOVHLQQ-UHFFFAOYSA-N
SMILES
COP(=O)(CC(=O)OCC1=CC=CC=C1)OC
Appearance (Color)
Clear colorless to light brown
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Refractive index
1.5050 to 1.5070 (20°C, 589 nm)
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text