Bismuth(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
Bismuth(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 387590250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS88189-03-1
IUPAC Namebismuth(3+) tritrifluoromethanesulfonate
Molecular FormulaC3BiF9O9S3
InChI KeyNYENCOMLZDQKNH-UHFFFAOYSA-K
SMILES[Bi+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Titration Complexometric31.0 to 33.0 % Bi
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only