Thermo Scientific Chemicals

Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals

Catalog number: 387600250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals

Catalog number: 387600250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 387600250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
82113-65-3
IUPAC Name
1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide
Molecular Formula
C2HF6NO4S2
InChI Key
ZXMGHDIOOHOAAE-UHFFFAOYSA-N
SMILES
FC(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
Appearance (Form)
Crystals
Titration with NaOH
>=98.5 %
Infrared spectrum
Conforms
Appearance (Color)
Colorless to brown
Melting point
50°C to 56°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text