Sodium chloride, pure, ca. 26% solution in water, Thermo Scientific Chemicals
Sodium chloride, pure, ca. 26% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium chloride, pure, ca. 26% solution in water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number 387640025
Price (USD)/ Each
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
Molecular Weight (g/mol)58.44
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless
Titration Argentometric25 to 28 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only