Thermo Scientific Chemicals

N-(2-Pyridyl)bis(trifluoromethanesulfonimide), 97%, Thermo Scientific Chemicals

Catalog number: 387650250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

N-(2-Pyridyl)bis(trifluoromethanesulfonimide), 97%, Thermo Scientific Chemicals

Catalog number: 387650250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 387650250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
145100-50-1
IUPAC Name
1,1,1-trifluoro-N-(pyridin-2-yl)-N-trifluoromethanesulfonylmethanesulfonamide
Molecular Formula
C7H4F6N2O4S2
InChI Key
DXLQEJHUQKKSRB-UHFFFAOYSA-N
SMILES
FC(F)(F)S(=O)(=O)N(C1=CC=CC=N1)S(=O)(=O)C(F)(F)F
Infrared spectrum
Conforms
Melting point
39°C to 43°C
Appearance (Color)
White to yellow
Appearance (Form)
Powder or crystals
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text