Potassium phosphate, tribasic, 97%, pure, anhydrous, Thermo Scientific Chemicals
Potassium phosphate, tribasic, 97%, pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium phosphate, tribasic, 97%, pure, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 387680250
Price (USD)/ Each
19.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
22.10 
Save 2.45 (11%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
19.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
22.10 
Save 2.45 (11%)
Add to cart
Potassium phosphate, tribasic, 97%, pure, anhydrous, Thermo Scientific Chemicals
Catalog number387680250
Price (USD)/ Each
19.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
22.10 
Save 2.45 (11%)
-
Add to cart
Chemical Identifiers
IUPAC Nametripotassium phosphate
Molecular FormulaK3O4P
InChI KeyLWIHDJKSTIGBAC-UHFFFAOYSA-K
SMILES[K+].[K+].[K+].[O-]P([O-])([O-])=O
Molecular Weight (g/mol)212.27
View more
SpecificationsSpecification SheetSpecification Sheet
Titration excess HCl>=96.0 %
Appearance (Color)White
Appearance (Form)Powder or granules and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO ā€“ Research Use Only
1x1 image pixel for data collection