Potassium vinyltrifluoroborate, 95%, Thermo Scientific Chemicals
Potassium vinyltrifluoroborate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium vinyltrifluoroborate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 387700050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namepotassium ethenyltrifluoroboranuide
Molecular FormulaC2H3BF3K
InChI KeyZCUMGICZWDOJEM-UHFFFAOYSA-N
SMILES[K+].F[B-](F)(F)C=C
Molecular Weight (g/mol)133.95
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Assay>=94 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only