Thermo Scientific Chemicals

Diphenyltetramethyldisilane, 97%, Thermo Scientific Chemicals

Catalog number: 387880010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Diphenyltetramethyldisilane, 97%, Thermo Scientific Chemicals

Catalog number: 387880010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 387880010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1145-98-8
IUPAC Name
1,1,2,2-tetramethyl-1,2-diphenyldisilane
Molecular Formula
C16H22Si2
InChI Key
IIOOIYHUTINYQO-UHFFFAOYSA-N
SMILES
C[Si](C)(C1=CC=CC=C1)[Si](C)(C)C1=CC=CC=C1
Appearance (Form)
Low melting mass
HPLC
>=96.0 %
Infrared spectrum
Conforms
Melting point
33°C to 38°C
Appearance (Color)
White to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text