Thermo Scientific Chemicals

5-Amino-1-pentanol, 50 wt.% aqueous solution, Thermo Scientific Chemicals

Catalog number: 387910250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

5-Amino-1-pentanol, 50 wt.% aqueous solution, Thermo Scientific Chemicals

Catalog number: 387910250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 387910250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2508-29-4
IUPAC Name
5-hydroxypentan-1-aminium
Molecular Formula
C5H14NO
InChI Key
LQGKDMHENBFVRC-UHFFFAOYSA-O
SMILES
[NH3+]CCCCCO
Infrared spectrum
Conforms
Appearance (Form)
Solution
Appearance (Color)
Clear colorless to light yellow
Titration with HCl
48.0 to 52.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text