D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP
Thermo Scientific Chemicals

D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP

CAS: 50-99-7 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Catalog number 388190250
Price (USD)
527.65
Online Exclusive
586.00
Save 58.35 (10%)
Each
-
Add to cart
Quantity:
25 kg
Request bulk or custom format
Price (USD)
527.65
Online Exclusive
586.00
Save 58.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS50-99-7
IUPAC Name6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular FormulaC6H12O6
InChI KeyWQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILESOCC1OC(O)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Specific optical rotation+52.5° to +53.3° (20°C, 589 nm) (C=10,H2O/dilute NH3)
Assay97.5 to 102.0 % (Anhydrous substance)
Related substances=<0.4 % (sum of impurities A and B)
Related substances=<0.15 % (impurity D)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link