Appearance (Color)Colorless to white to almost white
Appearance (Form)Crystalline powder or crystals
Identification APasses test
Identification BPasses test
Titration with NaOH99.5 to 101.0 % (on dried substance)