Thermo Scientific Chemicals

Potassium cyanide, 97+%, extra pure, Thermo Scientific Chemicals

Catalog number: 388310100
10 kg, Each
Thermo Scientific Chemicals

Potassium cyanide, 97+%, extra pure, Thermo Scientific Chemicals

Catalog number: 388310100
10 kg, Each
Quantity
Catalog number: 388310100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
151-50-8
IUPAC Name
potassium iminomethanide
Molecular Formula
CKN
InChI Key
NNFCIKHAZHQZJG-UHFFFAOYSA-N
SMILES
[K+].[C-]#N
Potassium formate
=<0.5 %
Titration Argentometric
>=97.0 %
Free alkali (KOH)
=<1 %
Appearance (Form)
Crystalline powder
Carbonate (CO3)
=<1 % (as K2CO3)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text