Potassium cyanide, 97+%, extra pure, Thermo Scientific Chemicals
Potassium cyanide, 97+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium cyanide, 97+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
10 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 388315000
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
IUPAC Namepotassium iminomethanide
Molecular FormulaCKN
InChI KeyNNFCIKHAZHQZJG-UHFFFAOYSA-N
SMILES[K+].[C-]#N
Molecular Weight (g/mol)65.12
View more
SpecificationsSpecification SheetSpecification Sheet
Potassium formate=<0.5 %
Titration Argentometric>=97.0 %
Free alkali (KOH)=<1 %
Appearance (Form)Crystalline powder
Carbonate (CO3)=<1 % (as K2CO3)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only