Benzyltrimethylammonium tribromide, 97%, Thermo Scientific Chemicals
Benzyltrimethylammonium tribromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyltrimethylammonium tribromide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 388320500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111865-47-5
IUPAC Namebenzyltrimethylazanium dibromine hydrobromide
Molecular FormulaC10H17Br3N
InChI KeyGSDDWZVWOXMLJZ-UHFFFAOYSA-N
SMILESBr.BrBr.C[N+](C)(C)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)Yellow to orange
Titration Iodometric>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only