Gold(I) chloride, 99+%, Thermo Scientific Chemicals
Gold(I) chloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold(I) chloride, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 388330050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namegold(1+) chloride
Molecular FormulaAuCl
InChI KeyFDWREHZXQUYJFJ-UHFFFAOYSA-M
SMILES[Cl-].[Au+]
Molecular Weight (g/mol)232.42
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow-green
Appearance (Form)Powder
Elemental Analysis>=99.0 % (ex Au)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only