Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 388381000
Price (USD)/ Each
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS124-40-3, 109-99-9
SpecificationsSpecification SheetSpecification Sheet
Strength1.8 to 2.2 M
Appearance (Form)Solution
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only