Chemical Identifiers
CAS112246-73-8
IUPAC Namechlorobis[(1S,2R,3S,5S)-2,6,6-trimethylbicyclo[3.1.1]heptan-3-yl]borane
Molecular FormulaC20H34BCl
InChI KeyPSEHHVRCDVOTID-NAVXHOJHSA-N
SMILESC[C@H]1[C@@H]2C[C@H](C[C@@H]1B(Cl)[C@H]1C[C@H]3C[C@@H]([C@@H]1C)C3(C)C)C2(C)C
View more