Thermo Scientific Chemicals

Phosphorus oxybromide, 95%, Thermo Scientific Chemicals

Catalog number: 388450250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Phosphorus oxybromide, 95%, Thermo Scientific Chemicals

Catalog number: 388450250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 388450250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-59-5
IUPAC Name
phosphoroyl tribromide
Molecular Formula
Br3OP
InChI Key
UXCDUFKZSUBXGM-UHFFFAOYSA-N
SMILES
BrP(Br)(Br)=O
Appearance (Form)
Crystals or fused mass
Appearance (Color)
Colorless to yellow to orange to light brown
Titration Argentometric
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text