Tin(IV) iodide, 99+%, -10 mesh, Thermo Scientific Chemicals
Tin(IV) iodide, 99+%, -10 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(IV) iodide, 99+%, -10 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 388560500
Price (USD)/ Each
576.65
Online exclusive
641.00 
Save 64.35 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
576.65
Online exclusive
641.00 
Save 64.35 (10%)
Add to cart
Tin(IV) iodide, 99+%, -10 mesh, Thermo Scientific Chemicals
Catalog number388560500
Price (USD)/ Each
576.65
Online exclusive
641.00 
Save 64.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7790-47-8
IUPAC Nametin(IV) iodide
Molecular FormulaH4I4Sn
InChI KeyZBCVLXKKLMNLGV-UHFFFAOYSA-N
SMILES[Sn].I.I.I.I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Brown
Titration Argentometric>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only