Potassium thiosulfate hydrate, 96+%, extra pure
Potassium thiosulfate hydrate, 96+%, extra pure
Potassium thiosulfate hydrate, 96+%, extra pure
Potassium thiosulfate hydrate, 96+%, extra pure
Thermo Scientific Chemicals

Potassium thiosulfate hydrate, 96+%, extra pure

CAS: 1264198-56-2 | K2O3S2 | 190.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 388691000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1264198-56-2
IUPAC Namedipotassium sulfanidesulfonate
Molecular FormulaK2O3S2
InChI KeyFGRVOLIFQGXPCT-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S([S-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodimetric96.0 to 104.0 % (On dry substance)
Appearance (Form)Powder or crystals and/or chunks
Loss on drying=<6 % (1 g, 150°C)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link