Thermo Scientific Chemicals

Allyltrimethoxysilane, 97+%, Thermo Scientific Chemicals

Catalog number: 388940050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

Allyltrimethoxysilane, 97+%, Thermo Scientific Chemicals

Catalog number: 388940050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 388940050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2551-83-9
IUPAC Name
trimethoxy(prop-2-en-1-yl)silane
Molecular Formula
C6H14O3Si
InChI Key
LFRDHGNFBLIJIY-UHFFFAOYSA-N
SMILES
CO[Si](CC=C)(OC)OC
Appearance (Form)
Liquid
Refractive index
1.4040 to 1.4060 (20°C, 589 nm)
Infrared spectrum
Conforms
GC
>=97.0 %
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text