Thermo Scientific Chemicals

Phosphoric acid, pure, Thermo Scientific Chemicals

Catalog number: 389020025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Phosphoric acid, pure, Thermo Scientific Chemicals

Catalog number: 389020025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 389020025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7664-38-2
IUPAC Name
phosphoric acid
Molecular Formula
H3O4P
InChI Key
NBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILES
OP(O)(O)=O
Appearance (Form)
Crystalline powder or crystals
Titration with NaOH
>=98.0 %
Iron (Fe)
=<10 ppm
Sulfate (SO4)
=<30 ppm
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text