Thermo Scientific Chemicals

n-Heptane, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 389170050
5 L, Each
Thermo Scientific Chemicals

n-Heptane, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 389170050
5 L, Each
Quantity
Catalog number: 389170050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
142-82-5
IUPAC Name
heptane
Molecular Formula
C7H16
InChI Key
IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES
CCCCCCC
GC
>=99.4 %
Bromine number
=<0.5
Copper (Cu)
=<0.02 ppm
Potassium (K)
=<0.1 ppm
Total sulfur (S)
=<0.001 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text