Thermo Scientific Chemicals

Calcium chloride hexahydrate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog number: 389260010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Calcium chloride hexahydrate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog number: 389260010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 389260010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7774-34-7
IUPAC Name
calcium hexahydrate dichloride
Molecular Formula
CaCl2H12O6
InChI Key
QHFQAJHNDKBRBO-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Ca++]
Appearance (Form)
Chunks
pH
4.5 to 8.5 (5 % solution)
Phosphate (PO4)
=<0.003 %
Ammonium (NH4)
=<0.01 %
Copper (Cu)
=<0.002 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text