Calcium chloride hexahydrate, 98+%, for analysis
Calcium chloride hexahydrate, 98+%, for analysis
Calcium chloride hexahydrate, 98+%, for analysis
Calcium chloride hexahydrate, 98+%, for analysis
Thermo Scientific Chemicals

Calcium chloride hexahydrate, 98+%, for analysis

CAS: 7774-34-7 | CaCl2·6H2O | 219.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 389262500
Price (USD)
25.65
Special offer
Online exclusive
Ends: 26-Sep-2025
31.20
Save 5.55 (18%)
Each
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
25.65
Special offer
Online exclusive
Ends: 26-Sep-2025
31.20
Save 5.55 (18%)
Each
Add to cart
Chemical Identifiers
CAS7774-34-7
IUPAC Namecalcium hexahydrate dichloride
Molecular FormulaCaCl2H12O6
InChI KeyQHFQAJHNDKBRBO-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Chunks
pH4.5 to 8.5 (5 % solution)
Phosphate (PO4)=<0.003 %
Ammonium (NH4)=<0.01 %
Copper (Cu)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link