Sodium cyclamate, 99%, Thermo Scientific Chemicals
Sodium cyclamate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium cyclamate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Plastic bottle
Catalog number 389282500
Price (USD)/ Each
70.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
83.20 
Save 12.55 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
70.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
83.20 
Save 12.55 (15%)
Add to cart
Sodium cyclamate, 99%, Thermo Scientific Chemicals
Catalog number389282500
Price (USD)/ Each
70.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
83.20 
Save 12.55 (15%)
-
Add to cart
Chemical Identifiers
CAS139-05-9
IUPAC Namesodium N-cyclohexylsulfamate
Molecular FormulaC6H12NNaO3S
InChI KeyUDIPTWFVPPPURJ-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)NC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HClO4>=98.5 % (On dry substance)
Loss on drying=<1 %
Sulfate (SO4)=<0.1 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only