Thermo Scientific Chemicals

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride, Thermo Scientific Chemicals

Catalog number: 389325000
500 g, Each
Thermo Scientific Chemicals

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride, Thermo Scientific Chemicals

Catalog number: 389325000
500 g, Each
Quantity
Catalog number: 389325000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25561-30-2
IUPAC Name
trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)ethanimidate
Molecular Formula
C8H18F3NOSi2
InChI Key
XCOBLONWWXQEBS-UHFFFAOYSA-N
SMILES
C[Si](C)(C)OC(=N[Si](C)(C)C)C(F)(F)F
GC
=<1.5 % TMCS
GC
>=98.0 % (excluding TMCS)
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text