Thermo Scientific Chemicals

Iron(II) chloride, 97%, anhydrous, Thermo Scientific Chemicals

Catalog number: 389352500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Iron(II) chloride, 97%, anhydrous, Thermo Scientific Chemicals

Catalog number: 389352500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 389352500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7758-94-3
IUPAC Name
λ²-iron(2+) dichloride
Molecular Formula
Cl2Fe
InChI Key
NMCUIPGRVMDVDB-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Fe++]
Titration with KMnO4
>=96.0 %
Appearance (Form)
Powder
Appearance (Color)
Off-white to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text