Thermo Scientific Chemicals

Water, Thermo Scientific Chemicals

Catalog number: 389390025
2.5 L, Plastic Bottle, Each
Thermo Scientific Chemicals

Water, Thermo Scientific Chemicals

Catalog number: 389390025
2.5 L, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 389390025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7732-18-5
IUPAC Name
water
Molecular Formula
H2O
InChI Key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES
O
Appearance (Color)
Clear colorless
Conductivity
=<1 µS/cm
Residue after evaporation
=<0.0003 %
UV
at 200 nm A: =<0.05
UV
at 210 nm A: =<0.009

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text