Water, for analysis, Thermo Scientific Chemicals
Water, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Water, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Plastic Bottle
Catalog number 389400025
Price (USD)/ Each
-
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS7732-18-5
IUPAC Namewater
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
pH5 to 8
Non-volatile matter=<0.0005 %
Phosphate (PO4)=<0.05 ppm
Silicate (as SiO2)=<0.01 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only