Thermo Scientific Chemicals

Tetrakis(hydroxymethyl)phosphonium chloride, ca. 80% solution in water, Thermo Scientific Chemicals

Catalog number: 389448000
800 mL, Glass bottle, Each
Thermo Scientific Chemicals

Tetrakis(hydroxymethyl)phosphonium chloride, ca. 80% solution in water, Thermo Scientific Chemicals

Catalog number: 389448000
800 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 389448000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
124-64-1, 7732-18-5
Infrared spectrum
Conforms
Appearance (Form)
Solution
Titration Argentometric
75 to 85 %
Appearance (Color)
Colorless to yellow-green

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text