Zinc chloride, 0.7M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Zinc chloride, 0.7M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc chloride, 0.7M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 389451000
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
Add to cart
Zinc chloride, 0.7M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Catalog number389451000
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7646-85-7, 109-99-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to hazy colorless
Water=<0.1 %
Appearance (Form)Solution
Titration Argentometric0.65 to 0.8 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only