Thermo Scientific Chemicals

Activated charcoal, iodinated, Thermo Scientific Chemicals

Catalog number: 389472500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Activated charcoal, iodinated, Thermo Scientific Chemicals

Catalog number: 389472500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 389472500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-44-0
IUPAC Name
carbon
Molecular Formula
C
InChI Key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES
[C]
Assay
5 % Exact strength on the CoA
Appearance (Color)
Black
Appearance (Form)
Rods

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text