Potassium polysulfide, pure
Potassium polysulfide, pure
Potassium polysulfide, pure
Potassium polysulfide, pure
Thermo Scientific Chemicals

Potassium polysulfide, pure

CAS: 37199-66-9 | (S)nK2 | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 389592500
Price (USD)
80.00
Each
Estimated availability date  Pending  
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
80.00
Each
Add to cart
Chemical Identifiers
CAS37199-66-9
Molecular Formula(S)nK2
SMILES[K+].[K+].*-[S--]-*
MDL NumberMFCD00148074
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Ochre to brown
Appearance (Form)Lumps
Titration Iodometric42.0 to 44.0 % (K2S)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0465770Certificate of AnalysisSep 10, 2024389590010, 389592500, 389590025
A0459765Certificate of AnalysisFeb 02, 2024389590010, 389592500, 389590025
A0457170Certificate of AnalysisNov 03, 2023389590010, 389592500, 389590025
A0452703Certificate of AnalysisMay 08, 2023389590010, 389592500, 389590025
A0447870Certificate of AnalysisJan 16, 2023389590010, 389592500, 389590025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection